2,4-Dichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

37947-0050 37947-0250
TS37947-0010EA 27.61 USD
TS37947-0010 TS37947-0050 TS37947-0250
2,4-Dichloropyridine 98%
2,4-Dichloropyridine
Formula: C₅H₃Cl₂N
Melting Pt: –1 °C
Density: 1.37 g/cm³
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00955618
CAS Number: 26452-80-2
EINECS: 247-717-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR