2,6-Dichloropyridine-N-oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloropyridine 1-oxide

37942-0010 37942-0050
TS37942-0010EA 32 USD
TS37942-0010 TS37942-0050
2,6-Dichloropyridine-N-oxide 98%
2,6-Dichloropyridine N-oxide
Formula: C₅H₃Cl₂NO
MW: 163.99 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00130246
CAS Number: 2587-00-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 134 to 142 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR