2-Iodopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

37933-0010 37933-0050
TS37933-0010EA 42 USD
TS37933-0010 TS37933-0050
2-Iodopyridine 98%
2-Iodopyridine
Formula: C₅H₄IN
Boiling Pt: 52 °C (.9 mmHg)
Melting Pt: 118…120 °C
Density: 1.928 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00464928
CAS Number: 5029-67-4

Order Now

Specification Test Results

Appearance Clear colorless to golden Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR