2-Chloro-5-iodopyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

37907-0010 37907-0050 37907-0250
TS37907-0010EA 50 USD
TS37907-0010 TS37907-0050 TS37907-0250
2-Chloro-5-iodopyridine 98+%
2-Chloro-5-iodopyridine
Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01863635
CAS Number: 69045-79-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 96 to 99 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR