2-Bromo-5-chloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37898-0010
37898-0050
TS37898-0010EA
92
USD
TS37898-0010
TS37898-0050
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Formula:
C₅H₃BrClN Melting Pt: 65…69 °C |
MDL Number:
MFCD00234006 CAS Number: 40473-01-6 UN: 2811 |
Specification Test Results
Appearance | Beige to yellow-brown Powder |
Infrared spectrum | Conforms |
Melting point | 65 to 69 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...