5-Bromo-2-fluoropyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

37846-0050 37846-0250
TS37846-0050EA 42 USD
TS37846-0050 TS37846-0250
5-Bromo-2-fluoropyridine 99%
5-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 63
Flash Pt: 73 °C
MDL Number: MFCD01863742
CAS Number: 766-11-0
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5330 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR