2-Amino-6-bromopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

37687-0050 37687-0250 37687-1000
TS37687-0050EA 73 USD
TS37687-0050 TS37687-0250 TS37687-1000
2-Amino-6-bromopyridine 98%
2-Amino-6-bromopyridine
Formula: C₅H₅BrN₂
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00137843
CAS Number: 19798-81-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR