3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DL-Lactide
37685-0250
37685-1000
TS37685-0250EA
85
USD
TS37685-0250
TS37685-1000
3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
3,6-Dimethyl-1,4-dioxane-2,5-dione
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 138…142 °C (8 mmHg) Melting Pt: 122…126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011685 CAS Number: 95-96-5 EINECS: 202-468-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 122 to 126 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...