5-(4-Chlorophenyl)-2-furoic acid 95+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37554-0010
37554-0050
TS37554-0010EA
53
USD
TS37554-0010
TS37554-0050
5-(4-Chlorophenyl)-2-furoic acid 95+%
5-(4-Chlorophenyl)-2-furoic acid
Formula:
C₁₁H₇ClO₃ Melting Pt: 198…201 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00216587 CAS Number: 41019-45-8 |
Specification Test Results
Appearance | Light brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...