1-Methyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

37354-0100 37354-0500
TS37354-0100EA 91 USD
TS37354-0100 TS37354-0500
1-Methyl-1H-pyrazole 98%
1-Methyl-1H-pyrazole
Formula: C₄H₆N₂
Boiling Pt: 127 °C (760 mmHg)
Density: 0.993 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00144943
CAS Number: 930-36-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR