4,4'-Dimethyl-2,2'-bipyridyl 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37323-0010
37323-0050
37323-0250
TS37323-0010EA
40
USD
TS37323-0010
TS37323-0050
TS37323-0250
4,4'-Dimethyl-2,2'-bipyridyl 99+%
4,4'-Dimethyl-2,2'-bipyridyl
Formula:
C₁₂H₁₂N₂ Melting Pt: 174…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006441 CAS Number: 1134-35-6 EINECS: 214-483-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Beige Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...