5,6-Epoxy-1-hexene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Epoxy-5-hexene
37258-0050
37258-0250
TS37258-0050EA
67
USD
TS37258-0050
TS37258-0250
5,6-Epoxy-1-hexene 98%
5,6-Epoxy-1-hexene
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 119…121 °C (1013 hPa) Melting Pt: –156 °C Density: 0.892 g/cm³ Flash Pt: 15 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010051 CAS Number: 10353-53-4 EINECS: 233-771-9 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4240 to 1.4260 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...