Piperonyloyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Benzodioxole-5-carbonyl chloride

37079-0010 37079-0100
TS37079-0010EA 44 USD
TS37079-0010 TS37079-0100
Piperonyloyl chloride 98%
Piperonyloyl chloride
Formula: C₈H₅ClO₃
MW: 184.575 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00016904
CAS Number: 25054-53-9
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 81 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR