3-Oxabicyclo[3.1.0]hexane-2,4-dione 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclopropane-1,2-dicarboxylic acid anhydride

37012-0010 37012-0050
TS37012-0010EA 40 USD
TS37012-0010 TS37012-0050
3-Oxabicyclo[3.1.0]hexane-2,4-dione 98%
3-Oxabicyclo[3.1.0]hexane-2,4-dione
Formula: C5H4O3
MW: 112.08 g/mol
Boiling Pt: 100…102 °C
Melting Pt: 54…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00126929
CAS Number: 5617-74-3

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 54 to 60 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR