2-Aminoimidazole hemisulfate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)

36886-0025 36886-0100
TS36886-0025EA 84 USD
TS36886-0025 TS36886-0100
2-Aminoimidazole hemisulfate 98+%
2-Aminoimidazole hemisulphate
Formula: C₆H₁₂N₆O₄S
MW: 264.26 g/mol
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013162
CAS Number: 1450-93-7
EINECS: 215-918-9

Order Now

Specification Test Results

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR