2-Chloro-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-4-picoline
36884-0050
36884-0250
36884-1000
TS36884-0050EA
88
USD
TS36884-0050
TS36884-0250
TS36884-1000
2-Chloro-4-methylpyridine 98%
2-Chloro-4-methylpyridine
Formula:
C₆H₆ClN MW: 127.57 g/mol Boiling Pt: 194…195 °C (760 mmHg) Density: 1.151 g/cm³ Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023418 CAS Number: 3678-62-4 EINECS: 222-951-2 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5280 to 1.5300 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...