N-Boc-L-prolinal 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-1-Boc-2-Formylpyrrolidine

36829-0010 36829-0050
TS36829-0010EA 144 USD
TS36829-0010 TS36829-0050 TS36829-0250
N-Boc-L-prolinal 97%
N-Boc-L-prolinal
Formula: C₁₀H₁₇NO₃
Boiling Pt: 211 °C (760 mmHg)
Density: 1.063 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00274186
CAS Number: 69610-41-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to orange Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4630 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -90° to -105° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR