1-Boc-4-hydroxypiperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate
36827-0050
36827-0250
36827-1000
TS36827-0050EA
52
USD
TS36827-0050
TS36827-0250
TS36827-1000
1-Boc-4-hydroxypiperidine 97%
1-Boc-4-hydroxypiperidine
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075174 CAS Number: 109384-19-2 |
Specification Test Results
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | 60 to 65 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...