1-(3-Aminopropyl)pyrrolidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(3-Aminopropyl)pyrrolidine, 3-(Pyrrolidin-1-yl)propan-1-amine

36809-0050 36809-0250
TS36809-0050EA 75 USD
TS36809-0050 TS36809-0250
1-(3-Aminopropyl)pyrrolidine 98%
1-(3-Aminopropyl)pyrrolidine
Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00014100
CAS Number: 23159-07-1
EINECS: 245-465-2
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR