(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(1-Ethylpyrrolidin-2-yl)methanamine, (S)-2-Aminomethyl-1-ethylpyrrolidine, (S)-1-ethyl-2-aminomethylpyrrolidine

36775-0010 36775-0050
TS36775-0010EA 50 USD
TS36775-0010 TS36775-0050
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 99%
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Density: 0.919 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00191371
CAS Number: 22795-99-9
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4660 to 1.4680 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -105° to -109° (20°C, 589 nm) (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR