3-Pyridineboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
36760-0010
36760-0050
TS36760-0010EA
61
USD
TS36760-0010
TS36760-0050
3-Pyridineboronic acid pinacol ester 97%
3-Pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₆BNO₂ MW: 205.06 g/mol Melting Pt: 103…108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03084758 CAS Number: 329214-79-1 |
Specification Test Results
Appearance | White to light brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 102 to 108 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...