3-Aminophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester
36759-0010
36759-0050
TS36759-0010EA
74
USD
TS36759-0010
TS36759-0050
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester
Formula:
C₁₂H₁₈BNO₂ Melting Pt: 89…94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03453668 CAS Number: 210907-84-9 |
Specification Test Results
Appearance | Light brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 89 to 94 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...