3-Carboxyphenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

36757-0010 36757-0050
TS36757-0010EA 35 USD
TS36757-0010 TS36757-0050
3-Carboxyphenylboronic acid pinacol ester 97%
3-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MW: 248.083 g/mol
Melting Pt: 206…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411930
CAS Number: 269409-73-6

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 206 to 211 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR