2-Aminophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester

36756-0010 36756-0050
TS36756-0010EA 76 USD
TS36756-0010 TS36756-0050
2-Aminophenylboronic acid pinacol ester 97%
2-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
Melting Pt: 64…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now

Specification Test Results

Appearance Cream Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 70 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR