4-Pyridineboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
36751-0010
36751-0050
TS36751-0010EA
94
USD
TS36751-0010
TS36751-0050
4-Pyridineboronic acid pinacol ester 97%
4-Pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₆BNO₂ Melting Pt: 153…155 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01319051 CAS Number: 181219-01-2 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...