1-Boc-piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

36728-0050 36728-0250
TS36728-0050EA 93 USD
TS36728-0050 TS36728-0250
1-Boc-piperazine 97%
1-Boc-piperazine
Formula: C₉H₁₈N₂O₂
Melting Pt: 43…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075265
CAS Number: 57260-71-6

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR