4-Amino-3,5-dichloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

36722-0050 36722-0250 36722-1000
TS36722-0050EA 28 USD
TS36722-0050 TS36722-0250 TS36722-1000
4-Amino-3,5-dichloropyridine 97%
4-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00125023
CAS Number: 22889-78-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 156 to 164 °C
Water ≤1 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR