1-(2-Tetrahydrofuroyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

36716-0010 36716-0050 36716-0250
TS36716-0010EA 30 USD
TS36716-0010 TS36716-0050 TS36716-0250
1-(2-Tetrahydrofuroyl)piperazine 98%
1-(2-Tetrahydrofuroyl)piperazine
Formula: C₉H₁₆N₂O₂
Boiling Pt: 140 °C (2mbar)
Density: 1.17 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00190247
CAS Number: 63074-07-7
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5195 to 1.5215 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR