2-(Piperazin-1-yl)pyrazine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(2-Pyrazinyl)piperazine

36713-0010 36713-0050
TS36713-0010EA 60 USD
TS36713-0010 TS36713-0050
2-(Piperazin-1-yl)pyrazine 98%
2-(Piperazin-1-yl)pyrazine
Formula: C₈H₁₂N₄
Boiling Pt: 105…108 °C (.5mbar)
Storage Temperature: Ambient
MDL Number: MFCD00040766
CAS Number: 34803-68-4
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR