4-Carboxyphenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

36639-0010 36639-0050
TS36639-0010EA 35 USD
TS36639-0010 TS36639-0050
4-Carboxyphenylboronic acid pinacol ester 97%
4-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MW: 248.08 g/mol
Melting Pt: 227…232 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 227 to 232 °C
Titration with NaOH 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR