6-Quinolinol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-6-ol, 6-Hydroxyquinoline

36493-0250 36493-1000
TS36493-0250EA 154 USD
TS36493-0250 TS36493-1000
6-Quinolinol 98%
6-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00047611
CAS Number: 580-16-5
EINECS: 209-454-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to grey to brown Powder
Infrared spectrum Conforms
Melting point 190 to 195 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR