3-Amino-6-chloro-4-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine

36099-0010 36099-0050
TS36099-0010EA 84 USD
TS36099-0010 TS36099-0050
3-Amino-6-chloro-4-methylpyridine 98%
3-Amino-6-chloro-4-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD03427661
CAS Number: 66909-38-4
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Bordeaux to brown Powder
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR