2-Chloro-6-methyl-5-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline

36082-0010 36082-0050
TS36082-0010EA 33 USD
TS36082-0010 TS36082-0050
2-Chloro-6-methyl-5-nitropyridine 98%
2-Chloro-6-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
MW: 172.57
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00234160
CAS Number: 22280-60-0
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Light beige to brown Powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR