3-Amino-2-chloro-6-methylpyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-2-chloro-6-picoline, 5-Amino-6-chloro-2-picoline

36074-0010 36074-0050
TS36074-0010EA 87 USD
TS36074-0010 TS36074-0050
3-Amino-2-chloro-6-methylpyridine 98+%
3-Amino-2-chloro-6-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Melting Pt: 74…80 °C
Storage Temperature: Ambient
MDL Number: MFCD03095213
CAS Number: 39745-40-9
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Cream to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR