2,5-Diaminopyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-2,5-diamine, Pyridine-2,5-diyldiamine

36069-0010 36069-0050 36069-0250
TS36069-0010EA 76 USD
TS36069-0010 TS36069-0050 TS36069-0250
2,5-Diaminopyridine 98+%
2,5-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 180 °C (12 mmHg)
Melting Pt: 110.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00129023
CAS Number: 4318-76-7
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow to purple Crystalline powder
Infrared spectrum Conforms
Melting point 106 to 110 °C
Additional info May contain up to about 10% water
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR