3-Bromo-2,5-diaminopyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromopyridine-2,5-diamine, 2,5-Diamino-3-bormopyridine, 3-Bromanylpyridine-2,5-diamine

36066-0010 36066-0050
TS36066-0010EA 83 USD
TS36066-0010 TS36066-0050
3-Bromo-2,5-diaminopyridine 99%
3-Bromo-2,5-diaminopyridine
Formula: C₅H₆BrN₃
MW: 188.028 g/mol
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD03427651
CAS Number: 896160-69-3

Order Now

Specification Test Results

Appearance Dark brown to violet Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR