3-Bromo-2-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

36065-0050 36065-0250
TS36065-0050EA 41 USD
TS36065-0050 TS36065-0250
3-Bromo-2-chloropyridine 98%
3-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 97 °C (1.3kPa)
Melting Pt: 57.9…58.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00234007
CAS Number: 52200-48-3

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals or flakes or solid
Infrared spectrum Conforms
Melting point 53 to 59 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR