4-Amino-3-bromopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

36062-0010 36062-0050
TS36062-0010EA 63 USD
TS36062-0010 TS36062-0050
4-Amino-3-bromopyridine 98%
4-Amino-3-bromopyridine
Formula: C₅H₅BrN₂
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD02068297
CAS Number: 13534-98-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 71 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR