2-Amino-3-bromopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36058-0050
36058-0250
TS36058-0050EA
48
USD
TS36058-0050
TS36058-0250
2-Amino-3-bromopyridine 98%
2-Amino-3-bromopyridine
Formula:
C₅H₅BrN₂ Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095187 CAS Number: 13534-99-1 |
Specification Test Results
Appearance | Grey to brown Powder |
Infrared spectrum | Conforms |
Melting point | 61 to 65 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...