3,4-(Methylenedioxy)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid

35958-0010 35958-0050
TS35958-0010EA 45 USD
TS35958-0010 TS35958-0050
3,4-(Methylenedioxy)phenylboronic acid 98%
3,4-(Methylenedioxy)phenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01009695
CAS Number: 94839-07-3

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Titration with NaOH 97.5 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR