5-Chloro-2-thiopheneboronic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-Chlorothiophen-2-yl)boronic acid, 5-Chloro-2-thienylboronic acid, 5-Chlorothiophene-2-boronic acid

35901-0010 35901-0050
TS35901-0010EA 34 USD
TS35901-0010 TS35901-0050
5-Chloro-2-thiopheneboronic acid 97+%
5-Chloro-2-thiopheneboronic acid
Formula: C₄H₄BClO₂S
MW: 162.4 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236030
CAS Number: 162607-18-3

Order Now

Specification Test Results

Appearance Off-white to light green to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with NaOH 97.0 to 103.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR