Bis(neopentyl glycolato)diboron 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
35889-0010
35889-0050
TS35889-0010EA
58
USD
TS35889-0010
TS35889-0050
Bis(neopentyl glycolato)diboron 98%
Bis(neopentyl glycolato)diboron
Formula:
C₁₀H₂₀B₂O₄ MW: 225.89 g/mol Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093062 CAS Number: 201733-56-4 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...