2-Bromo-5-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

35767-0010 35767-0050
TS35767-0010EA 34 USD
TS35767-0010 TS35767-0050
2-Bromo-5-nitropyridine 98%
2-Bromo-5-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Boiling Pt: 145…147 °C (10 mmHg)
Melting Pt: 137…141 °C
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR