5-Bromo-2-hydroxypyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol

35764-0050 35764-0250
TS35764-0050EA 56 USD
TS35764-0050 TS35764-0250
5-Bromo-2-hydroxypyridine 99%
5-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
Melting Pt: 179…182 °C
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now

Specification Test Results

Appearance Off-white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR