2-Phenylimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenyl-1H-imidazole

35643-0250 35643-1000
TS35643-0250EA 57 USD
TS35643-0250 TS35643-1000
2-Phenylimidazole 98%
2-Phenylimidazole
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 141…148 °C
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005186
CAS Number: 670-96-2
EINECS: 211-581-7

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline or granular powder
Infrared spectrum Conforms
Melting point 142 to 148 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR