6-Bromopiperonal 98%

Supplier: Thermo Scientific Chemicals
Warning

35592-0010 35592-0050 35592-0250
TS35592-0010EA 34 USD
TS35592-0010 TS35592-0050 TS35592-0250
6-Bromopiperonal 98%
6-Bromopiperonal
Formula: C₈H₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00022952
CAS Number: 15930-53-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR