Methyl-5-bromo-2-furoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35575-0010
35575-0050
35575-0250
TS35575-0010EA
45
USD
TS35575-0010
TS35575-0050
TS35575-0250
Methyl-5-bromo-2-furoate 98%
Methyl-5-bromo-2-furoate
Formula:
C₆H₅BrO₃ Melting Pt: 65…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092313 CAS Number: 2527-99-3 |
Specification Test Results
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...