4,4-Dimethyl-2-oxazoline 98%

Supplier: Thermo Scientific Chemicals
Danger

35527-0100 35527-0500
TS35527-0100EA 109 USD
TS35527-0100 TS35527-0500
4,4-Dimethyl-2-oxazoline 98%
4,4-Dimethyl-2-oxazoline
Formula: C₅H₉NO
Boiling Pt: 99…100 °C (760 mmHg)
Density: 0.94 g/cm³
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00014507
CAS Number: 30093-99-3
EINECS: 250-045-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4240 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR