2-Chloronicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile
35208-0010
35208-0050
35208-0250
TS35208-0010EA
27
USD
TS35208-0010
TS35208-0050
TS35208-0250
2-Chloronicotinonitrile 98%
2-Chloronicotinonitrile
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol Boiling Pt: 112 °C (1 mmHg) Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014628 CAS Number: 6602-54-6 EINECS: 229-550-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 105 to 110 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...