2-Bromo-4-methylpyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-picoline

35035-0010 35035-0100
TS35035-0010EA 32 USD
TS35035-0010 TS35035-0100
2-Bromo-4-methylpyridine 99%
2-Bromo-4-methylpyridine
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 87 °C (10 mmHg)
Melting Pt: 218…228 °C
Density: 1.545 g/cm³
Flash Pt: >109 °C
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5630 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR